Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_183387.2 | NP_899243.1 | 35 | 4863 | |
XM_006720070.3 | XP_006720133.1 | 33 | 4639 | ||
XM_011536528.2 | XP_011534830.1 | 35 | 4664 | ||
XM_011536530.2 | XP_011534832.1 | 34 | 4654 | ||
XM_011536531.2 | XP_011534833.1 | 33 | 4639 | ||
XM_011536532.2 | XP_011534834.1 | 35 | 4663 | ||
XM_011536533.2 | XP_011534835.1 | 33 | 4525 | ||
XM_011536534.2 | XP_011534836.1 | 36 | 4581 | ||
XM_011536536.2 | XP_011534838.1 | 35 | 4663 | ||
XM_017021062.1 | XP_016876551.1 | 35 | 4663 | ||
XM_017021063.1 | XP_016876552.1 | 34 | 4642 | ||
XM_017021064.1 | XP_016876553.1 | 35 | 4549 | ||
XM_017021065.1 | XP_016876554.1 | 32 | 4618 | ||
XM_017021066.1 | XP_016876555.1 | 34 | 4464 | ||
XM_017021067.1 | XP_016876556.1 | 35 | 4984 | ||
XM_017021068.1 | XP_016876557.1 | 34 | 4489 | ||
XM_017021069.1 | XP_016876558.1 | 35 | 4908 | ||
XM_017021070.1 | XP_016876559.1 | 34 | 4485 | ||
GenBank mRNA | AK127024.1 | - | 12 | 1333 | |
AK128086.1 | - | 26 | 3378 | ||
BC150639.1 | - | 35 | 4863 | 9057009 |