Gene Transcripts

Gene Symbol
SGMS2
Entrez Gene ID
166929
Gene Name
sphingomyelin synthase 2
Gene Aliases
SMS2
Chromosome Location
Chr. 4: 107823645 - 107915047 on Build GRCh38
UniGene ID
Hs.595423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136257.1 NP_001129729.1 2 540
NM_001136258.1 NP_001129730.1 3 732
NM_152621.5 NP_689834.1 2 816
XM_011531698.2 XP_011530000.1 3 870
XM_011531699.2 XP_011530001.1 4 744
XM_011531700.2 XP_011530002.1 3 641
XM_011531701.2 XP_011530003.1 4 1755
XM_011531702.2 XP_011530004.1 3 1683
XM_017007839.1 XP_016863328.1 3 874
XM_017007840.1 XP_016863329.1 4 996
XM_017007841.1 XP_016863330.1 3 870
GenBank mRNA AF452717.1 - 3 635
AK290344.1 - 3 734
AK314049.1 - 2 531
BC028705.1 - 2 541 4823252
BC041369.2 - 2 809 5273650
BX649142.1 - 1 197

View Full Product Details