Gene Transcripts
- Gene Symbol
- MYRFL
- Entrez Gene ID
- 196446
- Gene Name
- myelin regulatory factor-like
- Gene Aliases
- C12orf15, C12orf28, bcm1377
- Chromosome Location
- Chr. 12: 69824518 - 69958726 on Build GRCh38
- UniGene ID
- Hs.253773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_182530.2 |
NP_872336.2
|
24, 25 |
2851 |
|
XM_011539052.2 |
XP_011537354.1
|
23, 24 |
3332 |
|
XM_011539055.1 |
XP_011537357.1
|
22, 23 |
2319 |
|
XM_017018967.1 |
XP_016874456.1
|
23, 24 |
3133 |
|
XM_017018968.1 |
XP_016874457.1
|
24, 25 |
3587 |
|
XM_017018969.1 |
XP_016874458.1
|
23, 24 |
3067 |
|
XM_017018970.1 |
XP_016874459.1
|
22, 23 |
2877 |
|
XM_017018973.1 |
XP_016874462.1
|
20, 21 |
2548 |
|
GenBank mRNA |
AK057785.1 |
- |
11, 12 |
968 |
|
BC101820.1 |
- |
9, 10 |
803 |
|
BC143553.1 |
- |
8, 9 |
728 |
9052064 |
View Full Product Details