Gene Transcripts

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
Chromosome Location
Chr. 1: 153947672 - 153958656 on Build GRCh38
UniGene ID
Hs.406392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181715.2 NP_859066.1 13 1959
XM_005244946.1 XP_005245003.1 13 1955
XM_005244949.2 XP_005245006.1 13 1832
XM_017000575.1 XP_016856064.1 12 1728
XM_017000576.1 XP_016856065.1 12 1713
XM_017000577.1 XP_016856066.1 12 1857
XM_017000578.1 XP_016856067.1 12 1842
XM_017000579.1 XP_016856068.1 7 1254
GenBank mRNA AF289574.1 - 4 918
AK096382.1 - 11 1550
AK223559.1 - 13 1912
AY360172.1 - 13 1930
AY726597.1 - 13 1739
BC028886.1 - 2 194
BC033304.1 - 12 1628
BC053562.1 - 13 1870 6188068

View Full Product Details