Gene Transcripts

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
Chromosome Location
Chr. 5: 115962440 - 116027602 on Build GRCh38
UniGene ID
Hs.556880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173800.4 NP_776161.3 8 1703
XM_011543263.2 XP_011541565.1 8 1717
XM_011543264.2 XP_011541566.1 8 1717
XM_017009215.1 XP_016864704.1 8 1717
GenBank mRNA AK075131.1 - 3 238
AK291655.1 - 8 1717
AY560010.1 - 8 1591
BC036440.1 - 3 356
BC045809.1 - 3 331
BC060869.1 - 9 1630 30346049
BC068560.1 - 8 1703 30341970
BC070028.1 - 3 351
BC094716.1 - 3 351
BC109022.1 - 8 1567 40008501
BC109023.1 - 8 1567 40008502

View Full Product Details