Gene Transcripts

Gene Symbol
OR8K5
Entrez Gene ID
219453
Gene Name
olfactory receptor family 8 subfamily K member 5
Gene Aliases
OR11-174
Chromosome Location
Chr. 11: 56159394 - 56160317 on Build GRCh38
UniGene ID
Hs.553626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004058.2 NP_001004058.2 1 475

View Full Product Details