Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
219995
Gene Name
membrane spanning 4-domains A15
Gene Aliases
Chromosome Location
Chr. 11: 60756867 - 60776733 on Build GRCh38
UniGene ID
Hs.207465
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098835.1 NP_001092305.1 6 742
NM_001278242.1 NP_001265171.1 5 619
NM_152717.2 NP_689930.1 5 455
NR_103481.1 7 1029
XM_006718459.3 XP_006718522.1 5 462
XM_011544812.2 XP_011543114.1 6 706
XM_011544814.1 XP_011543116.1 6 571
GenBank mRNA AK091846.1 - 7 892
AK300233.1 - 5 533
AY584608.1 - 6 601
AY584609.1 - 5 478
BC031610.1 - 5 319 5176452
BI818815.1 - 6 607

View Full Product Details