Gene Transcripts

Gene Symbol
CFAP53
Entrez Gene ID
220136
Gene Name
cilia and flagella associated protein 53
Gene Aliases
CCDC11, HTX6
Chromosome Location
Chr. 18: 50227193 - 50266522 on Build GRCh38
UniGene ID
Hs.658630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145020.4 NP_659457.2 4 607
GenBank mRNA AK057305.1 - 3 328
AK302114.1 - 4 607
BC030606.2 - 4 580 4831081

View Full Product Details