Gene Transcripts

Gene Symbol
TIGD3
Entrez Gene ID
220359
Gene Name
tigger transposable element derived 3
Gene Aliases
Chromosome Location
Chr. 11: 65354811 - 65357613 on Build GRCh38
UniGene ID
Hs.632121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145719.2 NP_663771.1 2 237
GenBank mRNA AL558462.3 - 2 237
BC074862.2 - 1 95 30915228
DR002049.1 - 2 281

View Full Product Details