Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286635.1 | NP_001273564.1 | 4 | 490 | |
NM_152734.3 | NP_689947.2 | 3 | 390 | ||
XM_011514377.2 | XP_011512679.1 | 6 | 1253 | ||
XM_017010432.1 | XP_016865921.1 | 6 | 707 | ||
XM_017010433.1 | XP_016865922.1 | 6 | 708 | ||
XM_017010434.1 | XP_016865923.1 | 5 | 946 | ||
GenBank mRNA | AJ420498.1 | - | 3 | 397 | |
AK172780.1 | - | 3 | 299 | ||
AK300351.1 | - | 4 | 368 | ||
AY358637.1 | - | 4 | 364 | ||
BC023605.2 | - | 3 | 248 | ||
BC095423.1 | - | 4 | 403 | 30348810 | |
BC105946.1 | - | 3 | 297 | 40032607 | |
BC105947.1 | - | 3 | 298 | 40032604 | |
BC107489.1 | - | 3 | 298 | 40032608 | |
BX538108.1 | - | 3 | 283 | ||
BX647220.1 | - | 4 | 362 | ||
FR832871.1 | - | 4 | 434 | ||
FR832872.1 | - | 3 | 419 |