Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286355.1 | NP_001273284.1 | 3 | 1266 | |
NM_148963.3 | NP_683766.2 | 3 | 1266 | ||
XM_017010475.1 | XP_016865964.1 | 4 | 3199 | ||
XM_017010476.1 | XP_016865965.1 | 3 | 1227 | ||
GenBank mRNA | AF502962.1 | - | 3 | 1244 | |
AY435125.1 | - | 3 | 1255 | ||
AY435127.1 | - | 3 | 1255 |