Gene Transcripts

Gene Symbol
GPATCH11
Entrez Gene ID
253635
Gene Name
G-patch domain containing 11
Gene Aliases
CCDC75, CENP-Y, CENPY
Chromosome Location
Chr. 2: 37084451 - 37099244 on Build GRCh38
UniGene ID
Hs.595250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278505.1 NP_001265434.1 5 432
NM_001322249.1 NP_001309178.1 6 659
NM_174931.3 NP_777591.3 7 701
XM_017003748.1 XP_016859237.1 7 602
GenBank mRNA AK095667.1 - 5 365
AK289504.1 - 5 347
AK294697.1 - 7 615
BC071798.1 - 5 364
BM285380.1 - 7 701

View Full Product Details