Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098519.1 | NP_001091989.1 | 6 | 1076 | |
NM_152759.4 | NP_689972.3 | 6 | 965 | ||
XM_017019124.1 | XP_016874613.1 | 6 | 1084 | ||
GenBank mRNA | AK124107.1 | - | 8 | 1345 | |
BC029245.2 | - | 5 | 900 | 5169080 |