Gene Transcripts
- Gene Symbol
- MORN5
- Entrez Gene ID
- 254956
- Gene Name
- MORN repeat containing 5
- Gene Aliases
- C9orf113, C9orf18
- Chromosome Location
- Chr. 9: 122159661 - 122200088 on Build GRCh38
- UniGene ID
- Hs.71428
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198469.3 |
NP_940871.2
|
3, 4 |
379 |
|
XM_005251877.4 |
XP_005251934.1
|
3, 4 |
375 |
|
XM_005251878.3 |
XP_005251935.1
|
3, 4 |
350 |
|
XM_011518457.2 |
XP_011516759.1
|
3, 4 |
375 |
|
XM_011518458.1 |
XP_011516760.1
|
4, 5 |
458 |
|
GenBank mRNA |
AI026684.1 |
- |
2, 3 |
327 |
|
AI560944.1 |
- |
2, 3 |
345 |
|
AK128877.1 |
- |
3, 4 |
376 |
|
AK302081.1 |
- |
3, 4 |
353 |
|
BC133690.1 |
- |
3, 4 |
370 |
40119769 |
HY011325.1 |
- |
3, 4 |
380 |
|
View Full Product Details