Gene Transcripts

Gene Symbol
POC1B
Entrez Gene ID
282809
Gene Name
POC1 centriolar protein B
Gene Aliases
CORD20, PIX1, TUWD12, WDR51B
Chromosome Location
Chr. 12: 89419718 - 89526262 on Build GRCh38
UniGene ID
Hs.25130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199777.1 NP_001186706.1 1, 2 445
NM_172240.2 NP_758440.1 2, 3 464
GenBank mRNA AK074772.1 - 2, 3 202
AK123106.1 - 1, 2 442
BC026080.2 - 2, 3 226 4827689
DA851891.1 - 1, 2 445
EF011619.1 - 2, 3 226

View Full Product Details