Gene Transcripts

Gene Symbol
GOLGA6L2
Entrez Gene ID
283685
Gene Name
golgin A6 family-like 2
Gene Aliases
CT105
Chromosome Location
Chr. 15: 23439386 - 23447243 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304388.1 NP_001291317.1 4, 5 366
GenBank mRNA AK093463.1 - 4, 5 366
BC129820.1 - 4, 5 357 40113145
BC129821.1 - 3, 4 327

View Full Product Details