Gene Transcripts
- Gene Symbol
- NEK8
- Entrez Gene ID
- 284086
- Gene Name
- NIMA related kinase 8
- Gene Aliases
- JCK, NEK12A, NPHP9, RHPD2
- Chromosome Location
- Chr. 17: 28727899 - 28743457 on Build GRCh38
- UniGene ID
- Hs.448468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178170.2 |
NP_835464.1
|
12, 13 |
1739 |
|
XM_011524638.2 |
XP_011522940.1
|
13, 14 |
1982 |
|
XM_011524640.2 |
XP_011522942.1
|
13, 14 |
1943 |
|
XM_017024499.1 |
XP_016879988.1
|
12, 13 |
1788 |
|
XM_017024500.1 |
XP_016879989.1
|
12, 13 |
1824 |
|
GenBank mRNA |
AL833909.1 |
- |
3, 4 |
409 |
|
AY242354.1 |
- |
12, 13 |
1739 |
|
AY267371.1 |
- |
12, 13 |
1739 |
|
BC112240.1 |
- |
12, 13 |
1773 |
8327708 |
BC113705.1 |
- |
12, 13 |
1773 |
8322757 |
View Full Product Details