Gene Transcripts

Gene Symbol
PTRF
Entrez Gene ID
284119
Gene Name
polymerase I and transcript release factor
Gene Aliases
CAVIN, CAVIN1, CGL4, FKSG13, cavin-1
Chromosome Location
Chr. 17: 42402449 - 42423320 on Build GRCh38
UniGene ID
Hs.437191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012232.5 NP_036364.2 2 720
GenBank mRNA AF312393.1 - 2 917
AK000715.1 - 2 206
AK092406.1 - 3 521
AK298072.1 - 3 601
AK298563.1 - 3 575
AK314389.1 - 2 656
BC066123.1 - 2 656 5269172
BC073759.1 - 247 6572478

View Full Product Details