Gene Transcripts

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
Chromosome Location
Chr. 5: 41904313 - 41921636 on Build GRCh38
UniGene ID
Hs.29899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175921.5 NP_787117.3 5 572
XM_011514032.2 XP_011512334.1 6 701
GenBank mRNA BF439305.1 - 5 536

View Full Product Details