Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144877.2 | NP_001138349.1 | 16 | 1569 | |
NM_173690.4 | NP_775961.2 | 17 | 1638 | ||
GenBank mRNA | AF086497.1 | - | 3 | 183 | |
AK093983.1 | - | 16 | 1528 | ||
AK098024.1 | - | 17 | 1575 | ||
BC104030.2 | - | 17 | 1566 | 40025626 | |
BC104031.1 | - | 16 | 1497 | 40025627 |