Gene Transcripts
- Gene Symbol
- AGBL3
- Entrez Gene ID
- 340351
- Gene Name
- ATP/GTP binding protein like 3
- Gene Aliases
- CCP3
- Chromosome Location
- Chr. 7: 134985169 - 135135782 on Build GRCh38
- UniGene ID
- Hs.648616
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178563.3 |
NP_848658.3
|
10, 11 |
1984 |
|
XM_017012136.1 |
XP_016867625.1
|
10, 11 |
3306 |
|
XM_017012137.1 |
XP_016867626.1
|
10, 11 |
3588 |
|
XM_017012138.1 |
XP_016867627.1
|
9, 10 |
1790 |
|
XM_017012140.1 |
XP_016867629.1
|
9, 10 |
1790 |
|
XM_017012141.1 |
XP_016867630.1
|
9, 10 |
3123 |
|
XM_017012142.1 |
XP_016867631.1
|
9, 10 |
1790 |
|
XM_017012143.1 |
XP_016867632.1
|
9, 10 |
1790 |
|
GenBank mRNA |
AK302827.1 |
- |
10, 11 |
1984 |
|
BC030651.2 |
- |
10, 11 |
1959 |
5267615 |
View Full Product Details