Gene Transcripts

Gene Symbol
ITIH6
Entrez Gene ID
347365
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Gene Aliases
ITIH5L, UNQ6369, dJ14O9.1
Chromosome Location
Chr. X: 54748899 - 54798240 on Build GRCh38
UniGene ID
Hs.454272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198510.2 NP_940912.1 8 2456
GenBank mRNA AY358170.1 - 8 2456

View Full Product Details