Gene Transcripts

Gene Symbol
C2orf48
Entrez Gene ID
348738
Gene Name
chromosome 2 open reading frame 48
Gene Aliases
Chromosome Location
Chr. 2: 10141382 - 10211730 on Build GRCh38
UniGene ID
Hs.454981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182626.3 NP_872432.1 3, 4 490
GenBank mRNA AK057831.1 - 3, 4 490
BC093999.1 - 2, 3 232 7939844
BC094001.1 - 2, 3 231 7939846

View Full Product Details