Gene Transcripts

Gene Symbol
LRRC10
Entrez Gene ID
376132
Gene Name
leucine rich repeat containing 10
Gene Aliases
HRLRRP, LRRC10A
Chromosome Location
Chr. 12: 69608564 - 69611162 on Build GRCh38
UniGene ID
Hs.448708
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201550.3 NP_963844.2 1 421
GenBank mRNA AK123908.1 - 1 17
BC090881.1 - 1 421 30530394
BC106892.1 - 1 14
BC106893.1 - 1 17 40030470

View Full Product Details