Gene Transcripts

Gene Symbol
KCTD4
Entrez Gene ID
386618
Gene Name
potassium channel tetramerization domain containing 4
Gene Aliases
bA321C24.3
Chromosome Location
Chr. 13: 45192853 - 45201040 on Build GRCh38
UniGene ID
Hs.23406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198404.2 NP_940686.2 2 580
GenBank mRNA AK290675.1 - 2 582
BC018063.1 - 1 326 4814899

View Full Product Details