Gene Transcripts

Gene Symbol
OR51I1
Entrez Gene ID
390063
Gene Name
olfactory receptor family 51 subfamily I member 1
Gene Aliases
Chromosome Location
Chr. 11: 5440542 - 5441553 on Build GRCh38
UniGene ID
Hs.553732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005288.2 NP_001005288.1 1 415
GenBank mRNA BC151148.1 - 1 415

View Full Product Details