Gene Transcripts

Gene Symbol
OR5T1
Entrez Gene ID
390155
Gene Name
olfactory receptor family 5 subfamily T member 1
Gene Aliases
OR11-179, OR5T1P
Chromosome Location
Chr. 11: 56275639 - 56276619 on Build GRCh38
UniGene ID
Hs.553747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004745.1 NP_001004745.1 1 589
GenBank mRNA BC136992.1 - 1 675
BC136993.1 - 1 675 9020985

View Full Product Details