Gene Transcripts

Gene Symbol
OR6K3
Entrez Gene ID
391114
Gene Name
olfactory receptor family 6 subfamily K member 3
Gene Aliases
OR1-18
Chromosome Location
Chr. 1: 158717168 - 158718115 on Build GRCh38
UniGene ID
Hs.554488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005327.2 NP_001005327.2 1 571

View Full Product Details