Gene Transcripts

Gene Symbol
LGALS17A
Entrez Gene ID
400696
Gene Name
galectin 14 pseudogene
Gene Aliases
Chromosome Location
Chr. 19: 39679374 - 39686373 on Build GRCh38
UniGene ID
Hs.456377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_034156.1 4 487
GenBank mRNA AK123499.1 - 4 487
BC118595.1 - 2 180
BC122559.1 - 2 180
BC128606.1 - 3 391 40108149
FJ613353.1 - 4 441
FJ613354.1 - 4 468

View Full Product Details