Gene Transcripts

Gene Symbol
ZYG11A
Entrez Gene ID
440590
Gene Name
zyg-11 family member A, cell cycle regulator
Gene Aliases
ZYG11
Chromosome Location
Chr. 1: 52842511 - 52894575 on Build GRCh38
UniGene ID
Hs.658458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004339.2 NP_001004339.2 4 1435
NM_001307931.1 NP_001294860.1 3 683
XM_011541480.2 XP_011539782.1 4 1210
XM_011541481.2 XP_011539783.1 4 1210
XM_011541482.2 XP_011539784.1 4 1210
XM_011541483.2 XP_011539785.1 4 1210
XM_011541484.1 XP_011539786.1 4 1210
XM_011541485.1 XP_011539787.1 4 1210
XM_011541486.1 XP_011539788.1 4 1210
XM_011541487.2 XP_011539789.1 4 1210
XM_011541488.1 XP_011539790.1 4 1210
GenBank mRNA AY271826.1 - 3 674

View Full Product Details