Gene Transcripts
- Gene Symbol
- SUGT1P1
- Entrez Gene ID
- 441394
- Gene Name
- SGT1 homolog, MIS12 kinetochore complex assembly cochaperone pseudogene 1
- Gene Aliases
- SGT1P, SUGT1P, bA255A11.1
- Chromosome Location
- Chr. 9: 33500756 - 33511166 on Build GRCh38
- UniGene ID
- Hs.521441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003667.3 |
|
4 |
681 |
|
GenBank mRNA |
BE264023.1 |
- |
4 |
562 |
|
DA804368.1 |
- |
4 |
366 |
|
View Full Product Details