Gene Transcripts
- Gene Symbol
- ACTG1
- Entrez Gene ID
- 71
- Gene Name
- actin gamma 1
- Gene Aliases
- ACT, ACTG, BRWS2, DFNA20, DFNA26, HEL-176
- Chromosome Location
- Chr. 17: 81509971 - 81512866 on Build GRCh38
- UniGene ID
- Hs.514581
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199954.1 |
NP_001186883.1
|
3, 4 |
612 |
|
NM_001614.3 |
NP_001605.1
|
3, 4 |
493 |
|
NR_037688.1 |
|
3, 4 |
493 |
|
GenBank mRNA |
AK291937.1 |
- |
3, 4 |
426 |
|
AK301175.1 |
- |
3, 4 |
426 |
|
AK304637.1 |
- |
3, 4 |
423 |
|
BC000292.2 |
- |
3, 4 |
419 |
2819345 |
BC001920.1 |
- |
3, 4 |
382 |
2820489 |
BC004223.2 |
- |
2, 3 |
331 |
|
BC007442.2 |
- |
3, 4 |
413 |
2962953 |
BC009848.2 |
- |
3, 4 |
410 |
3939552 |
BC010999.2 |
- |
3, 4 |
412 |
3940861 |
BC012050.1 |
- |
3, 4 |
413 |
4554944 |
BC015005.1 |
- |
3, 4 |
408 |
4855463 |
BC015695.2 |
- |
3, 4 |
415 |
4842665 |
BC015779.1 |
- |
3, 4 |
389 |
4856294 |
BC017450.1 |
- |
2, 3 |
320 |
|
BC018774.1 |
- |
3, 4 |
413 |
4869628 |
BC023548.2 |
- |
1, 2 |
20 |
|
BC053572.1 |
- |
3, 4 |
406 |
3865767 |
BC063495.1 |
- |
2, 3 |
345 |
|
BT019856.1 |
- |
2, 3 |
354 |
|
X04098.1 |
- |
3, 4 |
427 |
|
View Full Product Details