Gene Transcripts

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
Chromosome Location
Chr. 2: 53970838 - 54305300 on Build GRCh38
UniGene ID
Hs.516173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320586.1 NP_001307515.1 7 688
NM_001320587.1 NP_001307516.1 6 595
NM_001320588.1 NP_001307517.1 4 389
NM_001320589.1 NP_001307518.1 5 710
NM_138448.3 NP_612457.1 4 640
XM_017005411.1 XP_016860900.1 8 1034
XM_017005412.1 XP_016860901.1 9 1104
XM_017005413.1 XP_016860902.1 9 1075
GenBank mRNA BC012290.1 - 4 409
BF671963.1 - 4 388
BG741807.1 - 6 589
BU543943.1 - 7 688
BU857820.1 - 5 568
CA308062.1 - 1 595
X84195.1 - 4 389

View Full Product Details