Gene Transcripts

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
Chromosome Location
Chr. 21: 44328944 - 44339417 on Build GRCh38
UniGene ID
Hs.517331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271440.1 NP_001258369.1 1 265
NM_001271441.1 NP_001258370.1 1 265
NM_004928.2 NP_004919.1 1 265
XM_006724053.3 XP_006724116.1 1 118
XM_017028470.1 XP_016883959.1 1 280
XM_017028471.1 XP_016883960.1 1 146
XM_017028472.1 XP_016883961.1 1 200
GenBank mRNA BC031300.1 - 1 256
BC072012.1 - 1 238 6645626
BM761336.1 - 1 265
U84569.1 - 1 237
Y11392.1 - 1 125
Z93322.1 - 1 64

View Full Product Details