Gene Transcripts

Gene Symbol
CYBA
Entrez Gene ID
1535
Gene Name
cytochrome b-245 alpha chain
Gene Aliases
p22-PHOX
Chromosome Location
Chr. 16: 88643289 - 88651084 on Build GRCh38
UniGene ID
Hs.513803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000101.3 NP_000092.2 4 287
XM_011522905.2 XP_011521207.1 4 282
GenBank mRNA BC006465.1 - 4 243
BT006861.1 - 4 216

View Full Product Details