Gene Transcripts

Gene Symbol
FOXC2
Entrez Gene ID
2303
Gene Name
forkhead box C2
Gene Aliases
FKHL14, LD, MFH-1, MFH1
Chromosome Location
Chr. 16: 86567251 - 86568933 on Build GRCh38
UniGene ID
Hs.436448
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005251.2 NP_005242.1 1 763
GenBank mRNA BC113437.1 - 1 763
BC113439.1 - 1 763 8322491

View Full Product Details