Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001312660.1 | NP_001299589.1 | 9 | 976 | |
NM_145870.2 | NP_665877.1 | 8 | 790 | ||
NM_145871.2 | NP_665878.2 | 7 | 664 | ||
XM_011536670.2 | XP_011534972.1 | 8 | 1391 | ||
XM_011536671.2 | XP_011534973.1 | 7 | 574 | ||
XM_017021243.1 | XP_016876732.1 | 7 | 1061 | ||
GenBank mRNA | AB209360.1 | - | 5 | 4916 | |
AJ001838.1 | - | 8 | 611 | ||
AK094344.1 | - | 1 | 1904 | ||
AK315609.1 | - | 8 | 634 | ||
BC001453.1 | - | 8 | 666 | 3139094 | |
BG330365.1 | - | 7 | 526 | ||
CR456987.1 | - | 7 | 508 | ||
U86529.1 | - | 8 | 611 |