Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021063.3 | NP_066407.1 | 1 | 121 | |
NM_138720.2 | NP_619790.1 | 1 | 121 | ||
XM_005249039.3 | XP_005249096.1 | 1 | 257 | ||
GenBank mRNA | AJ223353.1 | - | 1 | 100 | |
AK292712.1 | - | 1 | 117 | ||
BC002842.2 | - | 1 | 97 | 3659807 | |
BC096122.3 | - | 1 | 72 | 40002354 | |
BE245851.1 | - | 1 | 121 | ||
BM153714.1 | - | 1 | 121 |