Gene Transcripts

Gene Symbol
HBD
Entrez Gene ID
3045
Gene Name
hemoglobin subunit delta
Gene Aliases
Chromosome Location
Chr. 11: 5232829 - 5234628 on Build GRCh38
UniGene ID
Hs.699280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000519.3 NP_000510.1 3 751
GenBank mRNA BU664913.1 - 3 570

View Full Product Details