Gene Transcripts

Gene Symbol
MNX1
Entrez Gene ID
3110
Gene Name
motor neuron and pancreas homeobox 1
Gene Aliases
HB9, HLXB9, HOXHB9, SCRA1
Chromosome Location
Chr. 7: 157004853 - 157010653 on Build GRCh38
UniGene ID
Hs.37035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165255.1 NP_001158727.1 3 633
NM_005515.3 NP_005506.3 3 1189
GenBank mRNA AF107457.1 - 3 1073
X56537.1 - 3 396

View Full Product Details