Gene Transcripts

Gene Symbol
CEACAM6
Entrez Gene ID
4680
Gene Name
carcinoembryonic antigen related cell adhesion molecule 6
Gene Aliases
CD66c, CEAL, NCA
Chromosome Location
Chr. 19: 41755421 - 41772211 on Build GRCh38
UniGene ID
Hs.466814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002483.6 NP_002474.4 2 272
XM_011526990.1 XP_011525292.1 2 371
GenBank mRNA AK312542.1 - 2 257
BC005008.1 - 2 193 3640231
BT009774.1 - 2 153
DA442875.1 - 2 373
M18216.1 - 2 199
M18728.1 - 2 203
M29541.1 - 2 246

View Full Product Details