Gene Transcripts
- Gene Symbol
- REG3A
- Entrez Gene ID
- 5068
- Gene Name
- regenerating family member 3 alpha
- Gene Aliases
- HIP, HIP/PAP, INGAP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3
- Chromosome Location
- Chr. 2: 79157006 - 79159754 on Build GRCh38
- UniGene ID
- Hs.567312
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002580.2 |
NP_002571.1
|
4, 5 |
405 |
|
NM_138937.2 |
NP_620354.1
|
4, 5 |
382 |
|
NM_138938.2 |
NP_620355.1
|
3, 4 |
692 |
|
GenBank mRNA |
AK291973.1 |
- |
4, 5 |
381 |
|
AY544128.1 |
- |
3, 4 |
429 |
|
AY544130.1 |
- |
3, 4 |
375 |
|
BC036776.1 |
- |
3, 4 |
386 |
5749980 |
BQ777112.1 |
- |
4, 5 |
405 |
|
D13510.1 |
- |
3, 4 |
381 |
|
M84337.1 |
- |
3, 4 |
386 |
|
S51768.1 |
- |
3, 4 |
386 |
|
X68641.1 |
- |
3, 4 |
357 |
|
View Full Product Details