Gene Transcripts
- Gene Symbol
- PMS2P1
- Entrez Gene ID
- 5379
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 1
- Gene Aliases
- PMS2L1, PMS2L13, PMS2L6, PMS2L7, PMS2L8, PMS3, PMS8, PMSR1, PMSR2
- Chromosome Location
- Chr. 7: 100320640 - 100336307 on Build GRCh38
- UniGene ID
- Hs.634244
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003613.1 |
|
1 |
38 |
|
GenBank mRNA |
AB017004.1 |
- |
1 |
38 |
|
- Gene Symbol
- STAG3L1
- Entrez Gene ID
- 54441
- Gene Name
- stromal antigen 3-like 1 (pseudogene)
- Gene Aliases
- STAG3L1P, STAG3L2, STAG3L3
- Chromosome Location
- Chr. 7: 75359200 - 75367809 on Build GRCh38
- UniGene ID
- Hs.632310
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DC419304.1 |
- |
1 |
256 |
|
- Gene Symbol
- STAG3L2
- Entrez Gene ID
- 442582
- Gene Name
- stromal antigen 3-like 2 (pseudogene)
- Gene Aliases
- STAG3L1, STAG3L2P, STAG3L3
- Chromosome Location
- Chr. 7: 74881990 - 74890612 on Build GRCh38
- UniGene ID
- Hs.632310
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK307228.1 |
- |
2 |
225 |
|
- Gene Symbol
- STAG3L3
- Entrez Gene ID
- 442578
- Gene Name
- stromal antigen 3-like 3 (pseudogene)
- Gene Aliases
- STAG3L1, STAG3L2, STAG3L3P
- Chromosome Location
- Chr. 7: 72997295 - 73005943 on Build GRCh38
- UniGene ID
- Hs.632310
- Species
- Homo sapiens
- Gene Symbol
- STAG3L5P-PVRIG2P-PILRB
- Entrez Gene ID
- 101752399
- Gene Name
- STAG3L5P-PVRIG2P-PILRB readthrough
- Gene Aliases
- LOC101735302-LOC101752334-PILRB
- Chromosome Location
- Chr. 7: 100336065 - 100367831 on Build GRCh38
- UniGene ID
- Hs.521075
- Species
- Homo sapiens
View Full Product Details