Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001316307.1 | NP_001303236.1 | 5 | 507 | |
NM_005052.2 | NP_005043.1 | 5 | 507 | ||
GenBank mRNA | AF008591.1 | - | 5 | 471 | |
AF498966.1 | - | 5 | 401 | ||
BC009605.1 | - | 5 | 508 | 4284122 | |
BC015197.1 | - | 5 | 508 | 4275237 | |
BF941478.1 | - | 5 | 386 | ||
BI668056.1 | - | 5 | 461 | ||
BT019443.1 | - | 5 | 401 |