Gene Transcripts

Gene Symbol
SCT
Entrez Gene ID
6343
Gene Name
secretin
Gene Aliases
Chromosome Location
Chr. 11: 626095 - 627692 on Build GRCh38
UniGene ID
Hs.632324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021920.3 NP_068739.1 4 410
XM_017018109.1 XP_016873598.1 4 406
GenBank mRNA HY019819.1 - 4 410
HY243621.1 - 1 165

View Full Product Details