Gene Transcripts
- Gene Symbol
- AURKAPS1
- Entrez Gene ID
- 6791
- Gene Name
- aurora kinase A pseudogene 1
- Gene Aliases
- AurAps1, STK6P
- Chromosome Location
- Chr. 1: 220266179 - 220267715 on Build GRCh38
- UniGene ID
- Hs.654849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001587.1 |
|
1 |
186 |
|
View Full Product Details