Gene Transcripts

Gene Symbol
TBXA2R
Entrez Gene ID
6915
Gene Name
thromboxane A2 receptor
Gene Aliases
BDPLT13, TXA2-R
Chromosome Location
Chr. 19: 3594506 - 3608749 on Build GRCh38
UniGene ID
Hs.442530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001060.5 NP_001051.1 3 1188
NM_201636.2 NP_963998.2 3 1188
XM_011528214.2 XP_011526516.1 4 3222
GenBank mRNA AY429110.1 - 2 801
BC074749.2 - 2 840 30915459
BC074750.2 - 2 840 30915209

View Full Product Details