Gene Transcripts

Gene Symbol
TGFB1I1
Entrez Gene ID
7041
Gene Name
transforming growth factor beta 1 induced transcript 1
Gene Aliases
ARA55, HIC-5, HIC5, TSC-5
Chromosome Location
Chr. 16: 31472155 - 31477960 on Build GRCh38
UniGene ID
Hs.513530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042454.2 NP_001035919.1 11 1399
NM_001164719.1 NP_001158191.1 11 1368
NM_015927.4 NP_057011.2 11 1398
GenBank mRNA AB007836.1 - 10 1363
AF116343.1 - 10 1314
AK122975.1 - 6 4570
AK313327.1 - 11 1399
BC001507.3 - 11 1398 3030477
BC001830.2 - 11 1398 3030477
BC017288.2 - 11 1368 4901903
BC032545.1 - 10 1370 5535913

View Full Product Details