Gene Transcripts

Gene Symbol
ZNF28
Entrez Gene ID
7576
Gene Name
zinc finger protein 28
Gene Aliases
KOX24
Chromosome Location
Chr. 19: 52797408 - 52821686 on Build GRCh38
UniGene ID
Hs.732407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006969.3 NP_008900.3 4 996
NR_036599.1 5 1114
NR_036600.1 3 869
XM_006723354.3 XP_006723417.1 4 971
XM_011527262.2 XP_011525564.1 4 1026
GenBank mRNA AK131368.1 - 2 1594
AK131574.1 - 1 381
AK293074.1 - 3 869
AK304704.1 - 4 960
BC140720.1 - 3 897 9021588
BX640767.1 - 1 23
CR933598.1 - 4 959

View Full Product Details