Gene Transcripts

Gene Symbol
MBL1P
Entrez Gene ID
8512
Gene Name
mannose binding lectin 1, pseudogene
Gene Aliases
COLEC3P, MBL1P1
Chromosome Location
Chr. 10: 79920178 - 79923119 on Build GRCh38
UniGene ID
Hs.733579
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002724.2
3 1379
GenBank mRNA AF019382.1 - LIT2047
MBL1P1
3 1378

View Full Product Details